Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRSF Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | NRSF |
---|---|
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
NRSF Polyclonal specifically detects NRSF in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
NRSF | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box | |
Recombinant fusion protein containing a sequence corresponding to amino acids 760-1060 of human NRSF (NP_001180437.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Stem Cell Markers, Transcription Factors and Regulators | |
PBS with 50% glycerol, pH7.3. | |
5978 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title