Tocris™ NS 3763

Selective non-competitive kainate antagonist; selective for GLUK5

Manufacturer: Tocris™ 2187/50

 View more versions of this product

Catalog No. 21-875-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors NS 3763
    Purity >98
    Molecular Weight 404.38
    Quantity 50mg
    Product Type NS 3763

    Selective non-competitive kainate receptor antagonist; displays selectivity for GLUK5 subunit-containing receptors (IC50 values are 1.6 and > 30µM for homomeric GLUK5 and GLUK6 receptors respectively). Has minimal activity at AMPA and NMDA receptors (IC50> 30µM).

    • Chemical Name: 4,6-Bis(benzoylamino)-1,3-benzenedicarboxylic acid
    • Formula: C22H16N2O6
    • CAS Number: 70553-45-6
    • Solubility: Soluble to 25mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit