Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NS 3861

α3β2 full agonist; also α3β4 partial agonist

Manufacturer:  Tocris Bioscience™ 4831/50

 View more versions of this product

Catalog No. 483150


Add to cart
This item is not returnable. View return policy

Description

Description

α3β2 full agonist (EC50 = 1.6 μM). Also α3β4 partial agonist (EC50 = 1 μM). Exhibits minimal activity at α4β2 and α4β4. Displays high affinity for α3β4, α4β4, α3β2 and α4β2 (Ki values are 0.62, 7.8, 25 and 55 nM respectively).
Specifications

Specifications

3-(3-Bromo-2-thienyl)-8-methyl-8-azabicyclo[3.2.1]oct-2-ene fumarate
216853-60-0
400.29
Target: Additional Nicotinic Receptor Agonists
>98%
C12H14BrNS.C4H4O4
50mg
Safety and Handling

Safety and Handling

Recommended Storage : Store at RT

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit