Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NS 5806, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
KV4.3 channel activator; mediates the transient outward K+ current (Ito). Increases IKv4.3 peak current amplitude in CHO-K1 cells expressing KV4.3 and KChIP2 (EC50 = 5.3 μM). Inhibits KV1.4-mediated currents independently of KChIP2. Also slows the decay of KV4.2 and KV4.3 currents in the presence of KChIP2.
Specifications
Quantity | 10 mg |
Formula Weight | 574.07 |
Percent Purity | >99% |
Chemical Name or Material | NS 5806 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction