Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NS 6180, Tocris Bioscience™

Catalog Number 486410
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
353262-04-1
C16H12F3NOS
323.333
ZUIJXKLTUFCDGO-UHFFFAOYSA-N
4-3-trifluoromethyl benzyl-2h-benzo b 1,4 thiazin-3 4h-one, 4-3-trifluoromethyl phenyl methyl-1,4-benzothiazin-3-one, 4-3-trifluoromethyl phenyl methyl-2h-1,4-benzothiazin-3-one, 4-3-trifluoromethyl phenyl methyl-2h-1,4-benzothiazin-3 4h-one, 4-3-trifluoromethyl phenyl methyl-3,4-dihydro-2h-1,4-benzothiazin-3-one
11837221
4-[[3-(trifluoromethyl)phenyl]methyl]-1,4-benzothiazin-3-one
C1C(=O)N(C2=CC=CC=C2S1)CC3=CC(=CC=C3)C(F)(F)F
This item is not returnable. View return policy
353262-04-1
C16H12F3NOS
323.333
ZUIJXKLTUFCDGO-UHFFFAOYSA-N
4-3-trifluoromethyl benzyl-2h-benzo b 1,4 thiazin-3 4h-one, 4-3-trifluoromethyl phenyl methyl-1,4-benzothiazin-3-one, 4-3-trifluoromethyl phenyl methyl-2h-1,4-benzothiazin-3-one, 4-3-trifluoromethyl phenyl methyl-2h-1,4-benzothiazin-3 4h-one, 4-3-trifluoromethyl phenyl methyl-3,4-dihydro-2h-1,4-benzothiazin-3-one
11837221
4-[[3-(trifluoromethyl)phenyl]methyl]-1,4-benzothiazin-3-one
C1C(=O)N(C2=CC=CC=C2S1)CC3=CC(=CC=C3)C(F)(F)F

Potent KCa3.1 channel blocker

Potent KCa3.1 channel blocker (IC50 values are 9, 14 and 15 nM for rat, human and mouse erythrocyte KCa3.1 channels respectively). Exhibits ∽ 50% inhibition of KCa1.1, KV1.3, and KV11.1 channels, noradrenaline and dopamine transporters, L-type Ca2+ channels and melatonin receptors at a concentration of 10 μM. Potently inhibits IL-2 and IFN-γ production in rat lymphocytes (IC50 ∽ 50 nM); reduces DBNS-induced experimental colitis in rats.
Quantity 10 mg
Formula Weight 323.33
Percent Purity >99%
Chemical Name or Material NS 6180
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit