Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus NS5 Rabbit anti-Virus, DyLight 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242900UV

Catalog No. NBP242900UV


Add to cart

Description

Description

Dengue Virus NS5 Polyclonal antibody specifically detects Dengue Virus NS5 in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Dengue Virus NS5
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Rabbit
IgG
0.1 ml
Store at 4C in the dark.
Polyclonal
Virus
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified
The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
1494449
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only