Tocris™ NSC 146109 hydrochloride

Cell-permeable, genotype-selective antitumor agent; activates p53-dependent transcription

Manufacturer: Tocris™ 2185/50

 View more versions of this product

Catalog No. 21-855-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors NSC 146109 hydrochloride
    Purity >99
    Molecular Weight 316.85
    Quantity 50mg
    Product Type NSC 146109 hydrochloride

    Cell-permeable, genotype-selective antitumor agent that activates p53-dependent transcription. Increases levels of endogenous p53 in tumor cells and protects p53 from Mdm2-mediated degradation. Displays some selectivity for tumor cells vs. normal cells in an MTT cell viability assay.

    • Chemical Name: (10-Methyl-9-anthracenyl) methyl carbamimidothioic acid ester hydrochloride
    • Formula: C17H16N2S.HCl
    • CAS Number: 59474-01-0
    • Solubility: Soluble to 10mM in ethanol and to 100mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit