Learn More
Tocris Bioscience™ NSC 319726
Reactivator of mutant p53
Manufacturer:  Tocris Bioscience™ 5065/10
Catalog No. 50-651-0RD
Add to cart
This item is not returnable. View return policy
Description
Description
  • Restores wild type p53 structure and function to the p53R175 mutant
  • Inhibits growth of fibroblasts expressing the p53R175 mutation (IC50 = 8 nM)
  • Also inhibits growth of xenograft tumors expressing the p53R175 mutation in vivo
Specifications
Specifications
1-Azetidinecarbothioic acid [1-(2-pyridinyl)ethylidene]hydrazide
C11H14N4S
10 mg
71555-25-4
234.32
0.98
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C