Learn More
Tocris Bioscience™ NSC 319726 Available on GSA/VA Contract for Federal Government customers only.
Reactivator of mutant p53
Manufacturer:  Tocris Bioscience™ 5065/50
Catalog No. 50-655-0
Add to cart
This item is not returnable. View return policy
Description
Description
Reactivator of mutant p53. Restores wild type p53 structure and function to the p53R175 mutant. Inhibits growth of fibroblasts expressing the p53R175 mutation (IC50 = 8 nM). Also inhibits growth of xenograft tumors expressing the p53R175 mutation in vivo.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
NSC 319726
>98
50mg
NSC 319726
234.32