Tocris™ NSC 624206

Ubiquitin-activating enzyme (E1) inhibitor

Manufacturer: Tocris™ 4993/50

 View more versions of this product

Catalog No. 49-935-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors NSC 624206
    Purity >98
    Molecular Weight 410.51
    Quantity 50mg
    Product Type NSC 624206

    Ubiquitin-activating enzyme (E1) inhibitor; prevents tumour suppressor protein p27 ubiquitination in vitro. Blocks the ubiquitin-thioester formation step of the E1 activation reaction, but displays no effect on ubiquitin adenylation.

    • Chemical Name: N-[2-[[(4-Chlorophenyl)methyl]dithio]ethyl]-1-decanamine hydrochloride
    • Formula: C19H32ClNS2.HCl
    • CAS Number: 13116-77-3
    • Solubility: Soluble to 50mM in DMSO and to 20mM in ethanol
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit