Learn More
Tocris Bioscience NSC 624206
Ubiquitin-activating enzyme (E1) inhibitor
Manufacturer: Tocris Bioscience 4993/10
Description
Ubiquitin-activating enzyme (E1) inhibitor; prevents tumour suppressor protein p27 ubiquitination in vitro. Blocks the ubiquitin-thioester formation step of the E1 activation reaction, but displays no effect on ubiquitin adenylation.This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
NSC 624206 | |
>98 | |
10mg |
NSC 624206 | |
410.51 |