Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NSC 632839 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Inhibitor of ubiquitin isopeptidase activity that displays no effect on the proteolytic activity of the proteasome. Induces apoptosis via a Bcl-2-dependent and apoptosome-independent pathway of caspase activation (IC50 values are 15.65 and 16.23 μM in E1A and E1A/C9CN cells respectively).
Specifications
Quantity | 10 mg |
Formula Weight | 339.86 |
Percent Purity | >99% |
Chemical Name or Material | NSC 632839 hydrochloride |
Safety and Handling
Recommended Storage : Store at +4°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title