Learn More
Tocris Bioscience™ NSC 636819 Available on GSA/VA Contract for Federal Government customers only.
KDM4A/KDM4B inhibitor
$217.50 - $910.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
528710 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5287/10
Each for $217.50
Add to cart
528750 View Documents Tocris Bioscience™
5287/50
Each for $910.50
Add to cart
Description
Description
KDM4A/KDM4B inhibitor; inhibits histone demethylase activity on H3K9me3 and induces apoptosis in LNCaP prostate cancer cells in vitro. Also negatively regulates androgen-responsive genes.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.