Learn More
Alfa Aesar™ NSC 663284
Manufacturer:  Alfa Aesar™ J66520MA
 View more versions of this product
Catalog No. AAJ66520MA
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
10mg
-30°C to -10°C
383907-43-5
321.76g/mol
Light Sensitive
Red
Crystalline solid
C15H16ClN3O3
6-Chloro-7-(2-morpholin-4-yl-ethylamino)quinoline-5,8-dione; Cdc25 Phosphatase Inhibitor II