Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSC 66811, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
6964-62-1
C23H20N2O
340.426
WEENRMPCSWFMTE-UHFFFAOYSA-N
2-methyl-7-phenyl phenylamino methyl-8-quinolinol, 7-anilino phenyl methyl-2-methylquinolin-8-ol, 7-anilino phenyl-methyl-2-methyl-8-quinolinol, 2-methyl-7-phenyl phenylamino methyl quinolin-8-ol, 8-hydroxy-2-methyl-7-phenyl phenylamino methyl quinoline, d0c0tn, 7-anilino phenyl methyl-2-methyl-8-quinolinol, 2-methyl-7-phenyl-phenylamino methyl quinolin-8-ol, 8-quinolinol,2-methyl-7-phenyl phenylamino methyl
248986
7-[anilino(phenyl)methyl]-2-methylquinolin-8-ol
CC1=NC2=C(C=C1)C=CC(=C2O)C(C3=CC=CC=C3)NC4=CC=CC=C4
This item is not returnable. View return policy
6964-62-1
C23H20N2O
340.426
WEENRMPCSWFMTE-UHFFFAOYSA-N
2-methyl-7-phenyl phenylamino methyl-8-quinolinol, 7-anilino phenyl methyl-2-methylquinolin-8-ol, 7-anilino phenyl-methyl-2-methyl-8-quinolinol, 2-methyl-7-phenyl phenylamino methyl quinolin-8-ol, 8-hydroxy-2-methyl-7-phenyl phenylamino methyl quinoline, d0c0tn, 7-anilino phenyl methyl-2-methyl-8-quinolinol, 2-methyl-7-phenyl-phenylamino methyl quinolin-8-ol, 8-quinolinol,2-methyl-7-phenyl phenylamino methyl
248986
7-[anilino(phenyl)methyl]-2-methylquinolin-8-ol
CC1=NC2=C(C=C1)C=CC(=C2O)C(C3=CC=CC=C3)NC4=CC=CC=C4

MDM2 antagonist. Disrupts MDM2-p53 interaction

Potent MDM2 inhibitor (Ki = 120 nM) which disrupts MDM2-p53 interaction and activates p53 function. Induces p21, p53 and MDM2 accumulation in human colon cancer cells in vitro.
Quantity 10 mg
Formula Weight 340.42
Percent Purity >97%
Chemical Name or Material NSC 66811

Recommended Storage : Store at +4°C