Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NSC 693868, Tocris Bioscience™
Click to view available options
Quantity:
10mg
50mg
Description
Cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 0.4, 0.6, 4.4 - 5.6 μM for cdk5, cdk1 and cdk2, respectively). Also inhibits GSK-3 (IC50 = 1 uM).
Specifications
Quantity | 50mg |
Formula Weight | 185.19 |
Percent Purity | >99% |
Chemical Name or Material | NSC 693868 |
Safety and Handling
Recommended Storage : Store at +4°C