Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSC 697923, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
343351-67-7
C11H9NO5S
267.255
GAUHIPWCDXOLCZ-UHFFFAOYSA-N
2-4-methylphenyl sulfonyl-5-nitrofuran,2-nitro-5-tosylfuran,2-nitro-5-tosyl-furan,ncimech_000297,2-nitro-5-p-toluenesulfonylfuran,2-nitro-5-p-tolylsulfonyl furan,2-nitro-5-toluene-4-sulfonyl-furan,2-4-methylphenyl sulfonyl-5-nitro-furan,furan, 2-4-methylphenyl sulfonyl-5-nitro
394449
2-(4-methylphenyl)sulfonyl-5-nitrofuran
CC1=CC=C(C=C1)S(=O)(=O)C2=CC=C(O2)[N+](=O)[O-]
This item is not returnable. View return policy
343351-67-7
C11H9NO5S
267.255
GAUHIPWCDXOLCZ-UHFFFAOYSA-N
2-4-methylphenyl sulfonyl-5-nitrofuran,2-nitro-5-tosylfuran,2-nitro-5-tosyl-furan,ncimech_000297,2-nitro-5-p-toluenesulfonylfuran,2-nitro-5-p-tolylsulfonyl furan,2-nitro-5-toluene-4-sulfonyl-furan,2-4-methylphenyl sulfonyl-5-nitro-furan,furan, 2-4-methylphenyl sulfonyl-5-nitro
394449
2-(4-methylphenyl)sulfonyl-5-nitrofuran
CC1=CC=C(C=C1)S(=O)(=O)C2=CC=C(O2)[N+](=O)[O-]

Selective UBE2N inhibitor

Selective inhibitor of the E2 ubiquitin (Ub) conjugating enzyme UBE2N (Ubc13). Inhibits UBE2N-Ub conjugation and activity of the UBE2N-Ueva1A complex. Has no effect on UbcH5c activity. Inhibits NF-κB activation in diffuse large B-cell lymphoma (DLBCL) cells. Inhibits cell growth and induces apoptosis in DLBCL and neuroblastoma (NB) cell lines. Supresses NB cell xenograft tumor growth in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 267.26
Percent Purity >98%
Chemical Name or Material NSC 697923

Recommended Storage : Store at +4°C