Learn More
Aviva Systems Biology Corp NSDHL Antibody - middle region (ARP47199_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47199_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NSDHL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NSDHL. Alternate names: H105E3, XAP104, SDR31E1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59480