Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NSDHL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551417
Description
Immunogen sequence: LPYAMKPIDY YTETKILQER AVLGANDPEK NFLTTAIRPH GIFGPRDPQL VPILIEAARN GKMKFVIGNG KNLVDFTFVE NVVHGHILAA EQLSRDSTLG GKAFHITNDE PIPFWTFLSR I Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis. Mutations in this gene are associated with CHILD syndrome, which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis, and typically lethal in males. Alternatively spliced transcript variants with differing 5' UTR have been found for this gene.
Specifications
NSDHL | |
Polyclonal | |
Unconjugated | |
Nsdhl | |
AI747449; bare patches; Bpa; H105E3; NAD(P) dependent steroid dehydrogenase-like; NAD(P)-dependent steroid dehydrogenase-like protein; Nsdhl; Protein H105e3; SDR31E1; short chain dehydrogenase/reductase family 31E, member 1; sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating; Str; striated; XAP104 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18194, 309262, 50814 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15738, Q5PPL3, Q9R1J0 | |
Nsdhl | |
Recombinant protein corresponding to Human NSDHL. Recombinant protein control fragment (Product #RP-91901). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction