Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSDHL Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $299.00

Specifications

Antigen NSDHL
Immunogen Synthetic peptides corresponding to NSDHL(NAD(P) dependent steroid dehydrogenase-like) The peptide sequence was selected from the middle region of NSDHL. Peptide sequence RAVLGANDPEKNFLTTAIRPHGIFGPRDPQLVPILIEAARNGKMKFVIGN.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15979920 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15979920UL
20ul Each for $149.00
Add to cart
 
NBP159799 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159799
100ul Each for $299.00
Add to cart
 
Description

Description

NSDHL Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

NSDHL
Rabbit
Polyclonal
Unconjugated
50814
Purified
Western Blot
RUO
Synthetic peptides corresponding to NSDHL(NAD(P) dependent steroid dehydrogenase-like) The peptide sequence was selected from the middle region of NSDHL. Peptide sequence RAVLGANDPEKNFLTTAIRPHGIFGPRDPQLVPILIEAARNGKMKFVIGN.
Primary
IgG
EC 1.1.1.170, H105e3, member 1, NAD(P) dependent steroid dehydrogenase-like, SDR31E1, sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating, XAP104
PBS and 2% Sucrose with 0.09% Sodium Azide
Protein A purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit