Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nse2 Mouse anti-Human, FITC, Clone: OTI6A6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273070F
Description
Nse2 Monoclonal specifically detects Nse2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Nse2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
NSMCE2 | |
Full length recombinant protein of human Nse2 (NP_775956) produced in E.coli. | |
0.1 mL | |
286053.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI6A6 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
C8orf36, E3 SUMO-protein ligase NSE2, EC 6.3.2, EC 6.3.2.-, FLJ32440, hMMS21, methyl methanesulfonate sensitivity gene 21, MMS21 homolog, MMS21chromosome 8 open reading frame 36, Non-SMC element 2 homolog, non-SMC element 2 homolog (MMS21, S. cerevisiae), non-SMC element 2, MMS21 homolog (S. cerevisiae), Non-structural maintenance of chromosomes element 2 homolog, NSE2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title