Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSF Mouse anti-Bovine, Clone: NSF-1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266633

Catalog No. NBP266633


Add to cart

Description

Description

NSF Monoclonal antibody specifically detects NSF in Bovine samples. It is validated for Western Blot, Immunoprecipitation, Enzyme Immunoassay.
Specifications

Specifications

NSF
NSF-1
Western Blot, Immunoprecipitation 1:10 - 1:500, Enzyme Immunoassay
Affinity Purified
NSF
Recombinant human NSF
Affinity purified
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Bovine, Chicken, Human, Mouse, Rat
ELISA, Immunoprecipitation, Western Blot
Unconjugated
20MM sodium phosphate, 150MM sodium chloride, 50% glycerol, pH 7.5 with No Preservative
EC 3.6.4.6, NEM-sensitive fusion protein, N-ethylmaleimide-sensitive factor, N-ethylmaleimide-sensitive factor-like protein, N-ethylmaleimide-sensitive fusion protein, SKD2, vesicle-fusing ATPase, Vesicular-fusion protein NSF
Mouse
IgG1
100μg
Primary
4905
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit