Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAN Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548600

Catalog No. PA548600

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Couples the p55 TNF-receptor (TNF-R55 / TNFR1) to neutral sphingomyelinase (N-SMASE). Specifically binds to the N- smase activation domain of TNF-R55. May regulate ceramide production by N-SMASE.
Specifications

Specifications

FAN
Polyclonal
PBS with 0.09% sodium azide
Q92636, O35242
Protein FAN, Factor associated with neutral sphingomyelinase activation, Factor associated with N-SMase activation, NSMAF, FAN
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18201, 8439
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NSMAF
Liquid
NSMAF
KLH conjugated synthetic peptide between 28-57 amino acids from the N-terminal region of human NSMAF.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit