Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nsp9 Mouse anti-Virus, Alexa Fluor 350, Clone: 2C6.H1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221671AF350

Catalog No. NP221671A30


Add to cart

Description

Description

nsp9 Monoclonal antibody specifically detects nsp9 in Virus samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

nsp9
2C6.H1
Western Blot, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
nsp9
This antibody was produced in mice by repeated immunizations with E.coli derived full-length MHV-A59 nsp9 protein. This protein is part of the viral replicase polyprotein.
Protein A purified
Store at 4°C in the dark.
Monoclonal
No cross reactivity occurs with SARS CoV nsp9.
Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Non-structural protein 9, p12
Mouse
IgG2b Kappa
0.1 mL
Primary
1489749.0
Virus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit