Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NSUN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557561
Description
Immunogen sequence: PGGKSIALLQ CACPGYLHCN EYDSLRLRWL RQTLESFIPQ PLINVIKVSE LDGRKMGDAQ PEMFDKVLVD APCSNDRSWL FSSDSQKASC RISQR Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
NSUN3 may have S-adenosyl-L-methionine-dependent methyl-transferase activity.
Specifications
NSUN3 | |
Polyclonal | |
Unconjugated | |
NSUN3 | |
6720484A09Rik; AU022521; MST077; MSTP077; NOL1/NOP2/Sun domain family 3; NOL1/NOP2/Sun domain family member 3; NOL1/NOP2/Sun domain family member 3-like; NOL1/NOP2/Sun domain family, member 3; NOP2/Sun domain family, member 3; NOP2/Sun RNA methyltransferase 3; NOP2/Sun RNA methyltransferase family member 3; Nsun3; putative methyltransferase NSUN3; tRNA (cytosine(34)-C(5))-methyltransferase, mitochondrial; UG0651E06; Unknown (protein for MGC:142518) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
63899 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H649 | |
NSUN3 | |
Recombinant protein corresponding to Human NSUN3. Recombinant protein control fragment (Product #RP-96498). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction