Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSUN5B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Antigen NSUN5B
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123520
View Documents
Novus Biologicals
NBP30930925UL
25 μg N/A
NB123519
View Documents
Novus Biologicals
NBP309309100UL
100 μg N/A
Description

Description

NSUN5B Polyclonal antibody specifically detects NSUN5B in Human samples. It is validated for Western Blot
Specifications

Specifications

NSUN5B
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
155400
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Human
WBSCR20B
The immunogen is a synthetic peptide directed towards the N-terminal region of human NSUN5B. Peptide sequence GTPSPVRLHALAGFQQRALCHALTFPSLQRLVYSMCSLCQEENEDMVPDA
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit