Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT5C1A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549557

Catalog No. PA549557

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Dephosphorylates the 5 ft. and 2 ft.(3 ft.)-phosphates of deoxyribonucleotides and has a broad substrate specificity. Helps to regulate adenosine levels in heart during ischemia and hypoxia.
Specifications

Specifications

NT5C1A
Polyclonal
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9BXI3, A3KFX0
Cytosolic 5′-nucleotidase 1A, cN1A, Cytosolic 5′-nucleotidase IA, cN-I, cN-IA, NT5C1A
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
230718, 84618
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
NT5C1A
Liquid
NT5C1A
synthetic peptide corresponding to 165-192aa of human 5NT1A
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit