Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT5C3A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA124452
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA124452 50 μg
1 options

Catalog No. PIPA124452

Supplier: Thermo Scientific PA124452

Rabbit Polyclonal Antibody

PA1-24452 detects Pyrimidine 5-Prime-Nucleotidase (NT5C3) in Human samples. PA1-24452 has been successfully used in Western Blot procedures. PA1-24452 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human NT5C3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Pyrimidine 5-prime-nucleotidase (P5N), also called uridine 5-prime monophosphate hydrolase (UMPH), catalyzes the dephosphorylation of the pyrimidine 5-prime monophosphates UMP and CMP to the corresponding nucleosides. There are 2 isozymes of pyrimidine 5-prime nucleotidase in red blood cells, referred to as type I (UMPH1) and type II (UMPH2). The 2 enzymes are not separable by electrophoresis in humans but have distinct kinetic properties, and the proteins show no homology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT5C3A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene NT5C3A
Gene Accession No. Q9H0P0
Gene Alias NT5C3
Gene Symbols NT5C3A
Host Species Rabbit
Immunogen Polyclonal antibody from rabbits immunized with a synthetic peptide region of human NT5C3
Purification Method IgG fraction
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51251
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.