Learn More
NT5C3L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 162921AP
Catalog No. 50-173-0857
Add to cart
Description
Description
Specifically hydrolyzes 7-methylguanosine monophosphate (m(7)GMP) to 7-methylguanosine and inorganic phosphate. The specific activity for m(7)GMP may protect cells against undesired salvage of m(7)GMP and its incorporation into nucleic acids. Also has weak activity for CMP. UMP and purine nucleotides are poor substrates.
Specifications
Specifications
NT5C3L
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3UFY7, Q6AYP7, Q969T7
NT5C3L
Rabbit
IgG
150 μL
-20°C
Primary
115024, 360629, 68106
Western Blot
Unconjugated
NT5C3B
Liquid
Nt5c3b
NT5C3L Fusion Protein Ag9458
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat