Learn More
NT5D1 Mouse anti-Canine, Human, Non-human primate, Clone: OTI3A6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525210
Catalog No. PIMA525210
Add to cart
Description
Description
While the exact function of the protein encoded by this gene is not known, it belongs to the 5 ft.(3 ft.)-deoxyribonucleotidase family.
Specifications
Specifications
NT5D1
Monoclonal
0.78 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q5TFE4
C6orf200, LP2642, NT5C2L1, 5′-nucleotidase domain-containing protein 1, 5′-nucleotidase, cytosolic II-like 1 protein
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
221294, 475036, 716178
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3A6
Unconjugated
NT5DC1
Liquid
NT5DC1
Full length human recombit protein of NT5DC1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate