Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT5DC4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548195

Catalog No. PA548195

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

NT5DC4
Polyclonal
PBS with 0.09% sodium azide
Q86YG4
5′-nucleotidase domain-containing protein 4, NT5DC4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
284958
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NT5DC4
Liquid
NT5DC4
KLH conjugated synthetic peptide between 172-200 amino acids from the Central region of human NT5DC4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit