Sino Biological™ NT5E Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human NT5E (Met 1-Lys 547) , w/o propeptide, fused to polyhistidine tag at C-terminus

Manufacturer: Sino Biological™ 10904H08H50

 View more versions of this product

Catalog No. 10-904-H08H50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Structural Form Full Length, Recombinant
    Molecular Weight 59.2kd
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Source Human
    Endotoxin Concentration <1 EU/μg
    Common Name 5'-Nucleotidase, ECTO
    Gene Symbol NT5E
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family CD Proteins
    Expression System Mammalian
    Product Type NT5E Recombinant Human Protein
    For Use With (Application) Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 4907

    Expressed from DNA sequence encoding human NT5E (Met 1-Lys 547) , w/o propeptide, fused to polyhistidine tag at C-terminus

    • Measured by its ability to hydrolyze 5' phosphate group from substrate adenosine 5' monophosphate (AMP
    • Specific activity is >20,000 pmoles/min/µg
    • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit