Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ NT5E Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human NT5E (Met 1-Lys 547) , w/o propeptide, fused to polyhistidine tag at C-terminus

Manufacturer:  Sino Biological™ 10904H08H5

 View more versions of this product

Catalog No. 10-904-H08H5


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Expressed from DNA sequence encoding human NT5E (Met 1-Lys 547) , w/o propeptide, fused to polyhistidine tag at C-terminus

  • Measured by its ability to hydrolyze 5' phosphate group from substrate adenosine 5' monophosphate (AMP
  • Specific activity is >20,000 pmoles/min/µg
  • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Description & Specifications

Specifications

Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
4907
≥90% by SDS-PAGE
Human
<1 EU/μg
NT5E
CD Proteins
NT5E Recombinant Human Protein
Lyophilized
59.2kd
5μg
Full Length, Recombinant
5'-Nucleotidase, ECTO
Assay Standard
Mammalian
His Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit