Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT5M Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549543

Catalog No. PA549543

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Dephosphorylates specifically the 5 ft. and 2 ft.(3 ft.)- phosphates of uracil and thymine deoxyribonucleotides, and so protects mitochondrial DNA replication from excess dTTP. Has only marginal activity towards dIMP and dGMP.
Specifications

Specifications

NT5M
Polyclonal
PBS with 0.09% sodium azide
Q9NPB1
5′(3′)-deoxyribonucleotidase, mitochondrial, 5′, 3′-nucleotidase, mitochondrial, 3.1.3.-, Deoxy-5′-nucleotidase 2, dNT-2, NT5M, DNT2
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56953
Western Blot
Unconjugated
NT5M
Liquid
NT5M
KLH conjugated synthetic peptide between 195-226 amino acids from human NT5M.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit