Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NTE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552446
Description
Immunogen sequence: DVGYQYGKAV FGGWSRGNVI EKMLTDRRST DLNESRRADV LAFPSSGFTD LAEIVSRIEP PTSYVSDGCA DGEESDCLTE YEEDAGPDCS RDEGGSPEGA SPSTASEMEE EKSILRQRR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
This gene encodes a phospholipase that deacetylates intracellular phosphatidylcholine to produce glycerophosphocholine. It is thought to function in neurite outgrowth and process elongation during neuronal differentiation. The protein is anchored to the cytoplasmic face of the endoplasmic reticulum in both neurons and non-neuronal cells. Mutations in this gene result in autosomal recessive spastic paraplegia, and the protein is the target for neurodegeneration induced by organophosphorus compounds and chemical warfare agents. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
NTE | |
Polyclonal | |
Unconjugated | |
pnpla6 | |
5'-NT; 5'-nucleotidase; 5'-nucleotidase ecto; 5'-nucleotidase, ecto (CD73); AI661849; BNHS; CALJA; CD73; E5NT; ecto-5'-nucleotidase; eN; eNT; iPLA2delta; LNMS; MSws; Neuropathy target esterase; NT; NT5; NT5E; NTE; NTEMND; OMCS; patatin like phospholipase domain containing 6; patatin-like phospholipase domain containing 6; patatin-like phospholipase domain-containing protein 6; PNPLA6; Purine 5-Prime-Nucleotidase; RGD1564611; SPG39; Swiss cheese; Swiss-cheese; sws; wu:fy20f06 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10908 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IY17 | |
pnpla6 | |
Recombinant protein corresponding to Human NTE. Recombinant protein control fragment (Product #RP-89959). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction