Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NTMT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550449
Description
The antibody detects endogenous levels of total METTL11A / NTM1A protein.
The METTL11A gene encodes an N-terminal methyltransferase for the RAN (MIM 601179) guanine nucleotide exchange factor regulator of chromosome condensation 1 (RCC1; MIM 179710). METTL11A enzyme alpha-N-methylates other protein targets such as SET (MIM 600960) and RB (MIM 180200).
Specifications
NTMT1 | |
Polyclonal | |
Unconjugated | |
NTMT1 | |
2610205E22Rik; AD-003; AL033331; AL033332; Alpha N-terminal protein methyltransferase 1A; C9orf32; HOMT1A; methyltransferase like 11A; methyltransferase-like protein 11A; METTL11A; NRMT; NRMT1; N-terminal RCC1 methyltransferase; N-terminal Xaa-Pro-Lys N-methyltransferase 1; N-terminal Xaa-Pro-Lys N-methyltransferase 1, N-terminally processed; NTM1A; NTMT1; RGD1306582; X-Pro-Lys N-terminal protein methyltransferase 1A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28989 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9BV86 | |
NTMT1 | |
Full length fusion protein of Human NTMT1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction