Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTN3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548722

Catalog No. PA548722


Add to cart

Description

Description

Netrins control guidance of CNS commissural axons and peripheral motor axons.
Specifications

Specifications

NTN3
Polyclonal
PBS with 0.09% sodium azide
O00634
Netrin-3, Netrin-2-like protein, NTN3, NTN2L
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4917
Western Blot
Unconjugated
NTN3
Liquid
NTN3
KLH conjugated synthetic peptide between 431-459 amino acids from the C-terminal region of human NTN3.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit