Tocris™ NTNCB hydrochloride

Potent and selective non-peptidic NPY Y5 antagonist

Manufacturer: Tocris™ 2155/50

 View more versions of this product

Catalog No. 21-555-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors NTNCB hydrochloride
    Purity >99
    Molecular Weight 508.07
    Quantity 50mg
    Product Type NTNCB hydrochloride

    Selective, non-peptide competitive NPY Y5 antagonist. Ki values are 8.0 and 16032nM at human recombinant Y5 and Y1 receptors respectively. Potently antagonizes NPY inhibition of forskolin-stimulated cAMP. Displays some affinity for cloned human D2 and α2C receptors (Ki values are 63 and 100nM respectively).

    • Chemical Name: 2-Nitro-N-[[trans-4-[[[(1,2,3,4-tetrahydro-2-naphthalenyl)methyl]amino]methyl]cyclohexyl]methyl]benzenesulfonamide hydrochloride
    • Formula: C25H33N3O4S.HCl
    • CAS Number: 191931-56-3
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit