Learn More
NTNG1 Rat anti-Mouse, Alexa Fluor 700, Clone: RM0292-4K62, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211810AF700
Catalog No. NB211810700
Add to cart
Description
Description
NTNG1 Monoclonal specifically detects NTNG1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
NTNG1
RM0292-4K62
Western Blot
Purified
NTNG1
Mouse NTNG1
Protein G purified
Neuroscience
Primary
22854
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
axon guidance molecule, KIAA0976YLSR571, laminet 1, laminet-1, Lmnt1, netrin G1, netrin G1f, netrin-G1
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse