Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Human NTRK2 (TRKB), His Tag Recombinant Protein
Human NTRK2 (TRKB), His Tag Recombinant Protein
Supplier: Thermo Scientific™ PV3616
Description
NTRK2 (TRKB) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. NTRK2 is the receptor for brain-derived neurotrophic factor (BDNF). Together, NTRK2 and BDNF regulate both short-term synaptic functions and long-term potentiation of brain synapses.Specifications
NP_006171 | |
4915 | |
10 μg | |
Neurotrophic Receptor (NTRK) Kinase Family | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity | |
Dry Ice | |
NTRK2 | |
Baculovirus | |
Recombinant, Cytoplasmic | |
Receptor Tyrosine Kinases |
Kinase Assay | |
40.9 kDa | |
NTRK2 (TRKB) | |
aa 526-838 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 300 mM NaCl, 0.5 mM EDTA, 0.05% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
neurotrophic tyrosine kinase, receptor, type 2 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction