Learn More
NTRK2 (TRKB) Recombinant Human Protein
$450.00
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
PV3616 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
PV3616
N/A 10μg
Each for $450.00
Add to cart
Description
Description
NTRK2 (TRKB) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. NTRK2 is the receptor for brain-derived neurotrophic factor (BDNF). Together, NTRK2 and BDNF regulate both short-term synaptic functions and long-term potentiation of brain synapses.