Learn More
Aviva Systems Biology Corp NTRK3 Antibody - C-terminal region (ARP51318_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51318_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NTRK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NTRK3. Alternate names: TRKC, gp145(trkC)ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60543