Learn More
NTSR1 Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Supplier:  Thermo Scientific PA130157
Catalog No. PIPA130157
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-30157 detects NTS1 from rat samples. PA1-30157 has been successfully used in immunocytochemistry applications. The PA1-30157 immunogen is: TVGTHNGLEHSTFNMTIEPGRVQAYC Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NTSR1
Polyclonal
Ntsr1
NTSR1
TVGTHNGLEHSTFNMTIEPGRVQAYC
RUO
366274
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Unconjugated
P20789
Guinea Pig
50 μL
Primary
Rat
Antibody
IgG
Recommendations
Recommendations