NU 1025, Tocris

Potent PARP inhibitor

Manufacturer: R&D Systems 1401/50

Quantity 50mg

 View more versions of this product

Catalog No. 14-015-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C9H8N2O2
    Formula Weight 176.17
    Synonym 8-Hydroxy-2-methyl-4(3H)-quinazolinone
    Chemical Name or Material NU 1025

    Specifications

    Formula Weight 176.17
    Quantity 50mg
    Assay Percent Range >99%
    Molecular Formula C9H8N2O2
    CAS 90417-38-2
    Synonym 8-Hydroxy-2-methyl-4(3H)-quinazolinone
    Recommended Storage Store at Room Temperature
    Chemical Name or Material NU 1025

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit