Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUBPL Mouse anti-Human, Clone: OTI1C5, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503726
Description
Required for the assembly of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). May deliver of one or more Fe-S clusters to complex I subunits.Specifications
NUBPL | |
Monoclonal | |
0.61 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8TB37 | |
C14orf127, IND1, huInd1 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
80224 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1C5 | |
Unconjugated | |
NUBPL | |
Liquid | |
NUBPL | |
Human recombit protein fragment corresponding to amino acids 1-250 of human NUBPL produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title