Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUBPL Mouse anti-Human, Clone: OTI2C8, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503725

Catalog No. 50-167-4549


Add to cart

Description

Description

Required for the assembly of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). May deliver of one or more Fe-S clusters to complex I subunits.
Specifications

Specifications

NUBPL
Monoclonal
0.68 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8TB37
C14orf127, IND1, huInd1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80224
Flow Cytometry, Western Blot
OTI2C8
Unconjugated
NUBPL
Liquid
NUBPL
Human recombit protein fragment corresponding to amino acids 1-250 of human NUBPL produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit