Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUBPL Monoclonal Antibody (OTI5C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA503748
Description
Required for the assembly of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). May deliver of one or more Fe-S clusters to complex I subunits.Specifications
NUBPL | |
Monoclonal | |
0.67 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8TB37 | |
NUBPL | |
Human recombit protein fragment corresponding to amino acids 1-250 of human NUBPL produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI5C12 | |
Unconjugated | |
NUBPL | |
C14orf127, IND1, huInd1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
80224 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction