Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nuclear Antigen Rabbit anti-Human, Mouse, Rat, Clone: NM2984R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Nuclear Antigen
Clone NM2984R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 0.25-0.5 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122079
View Documents
Novus Biologicals
NBP30730310UG
100 μg
Each for $406.52
Add to cart
 
NB122080
View Documents
Novus Biologicals
NBP30730320UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Nuclear Antigen
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
IgG
Primary
Store at 4°C.
NM2984R
Immunohistochemistry-Paraffin 0.25-0.5 ug/ml
Unconjugated
Rabbit
Human, Mouse, Rat
Nuclei of HL60 cells
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit