Learn More
R&D Systems™ Nuclear Fast Red
R&D Systems™ Nuclear Fast Red is the ideal counterstain for kits employing the TACS Blue Label™ chromogenic substrate.
Manufacturer:  R&D Systems™ 48003017
Catalog No. 48-003-017
Add to cart
This item is not returnable. View return policy
Description
Description
  • Hazard Statements (GHS-US): H318 - Causes serious eye damage
  • Precautionary Statements (GHS-US):
    • P280 - Wear protective gloves, protective clothing, and eye protection
    • P305+P351+P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P310 - Immediately call a poison center or doctor.
  • WHMIS Classification: Class E - Corrosive Material
Specifications
Specifications
50mL
Aluminum sulfate hydrate - (CAS No) 17927-65-0
Store the unopened product at room temperature