Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleobindin 1 Mouse anti-Human, Mouse, Rat, FITC, Clone: OTI1A5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273086F
Description
Nucleobindin 1 Monoclonal specifically detects Nucleobindin 1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Nucleobindin 1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
NUCB1 | |
Full length recombinant protein of human Nucleobindin 1 (NP_006175) produced in HEK293T cells. | |
0.1 mL | |
4924.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1A5 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DKFZp686A15286, FLJ40471, NUCCALNUC, nucleobindin 1, nucleobindin-1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title